3-(4-Methyl-4H-[1,2,4]triazol-3-yl)phenylamine 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-(4-Methyl-4H-1,2,4-triazol-3-yl)aniline
OR12331-1G
OR12331-5G
OR12331-10G
APOSOR12331-1GEA
429
EUR
APOSOR12331-1G
APOSOR12331-5G
APOSOR12331-10G
3-(4-Methyl-4H-[1,2,4]triazol-3-yl)phenylamine 97%
3-(4-Methyl-4H-[1,2,4]triazol-3-yl)phenylamine
Formula:
C₉H₁₀N₄ MW: 174,21 g/mol Melting Pt: 158…159 °C Storage Temperature: Ambient |
MDL Number:
MFCD02853082 CAS Number: 252928-74-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...