4-Hydroxychalcone 97%

Supplier: Apollo Scientific
Warning

OR1234-25G OR1234-5G
APOSOR1234-25GEA 241 EUR
APOSOR1234-25G APOSOR1234-5G
4-Hydroxychalcone 97%
4-Hydroxychalcone
Formula: C₁₅H₁₂O₂
Melting Pt: 180…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00016488
CAS Number: 20426-12-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR