3-Amino-5-hydroxybenzoic acid hydrochloride 95%

Supplier: Apollo Scientific

OR12368-1G OR12368-5G
APOSOR12368-1GEA 56.8 EUR
APOSOR12368-1G APOSOR12368-5G
3-Amino-5-hydroxybenzoic acid hydrochloride 95%
3-Amino-5-hydroxybenzoic acid hydrochloride
Formula: C₇H₈ClNO₃
MW: 189,6000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00043420
CAS Number: 14206-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR