(5-Pyrrolidin-1-ylpyridin-2-yl)methanol 97%
Supplier: Apollo Scientific
|
Warning
|
OR12405-1G
OR12405-5G
APOSOR12405-1GEA
0
EUR
APOSOR12405-1G
APOSOR12405-5G
(5-Pyrrolidin-1-ylpyridin-2-yl)methanol 97%
(5-Pyrrolidin-1-ylpyridin-2-yl)methanol
Formula:
C₁₀H₁₄N₂O MW: 178,24 g/mol Melting Pt: 133,5…136 °C Storage Temperature: Ambient |
MDL Number:
MFCD09702371 CAS Number: 930110-98-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...