3-Methyl-5-isoxazolecarboxylic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Methylisoxazole-5-carboxylic acid
OR12419-1G
OR12419-250MG
APOSOR12419-1GEA
29
EUR
APOSOR12419-1G
APOSOR12419-250MG
3-Methyl-5-isoxazolecarboxylic acid
3-Methyl-5-isoxazolecarboxylic acid
Formula:
C₅H₅NO₃ MW: 127,10 g/mol Melting Pt: 210…213 °C Storage Temperature: Ambient |
MDL Number:
MFCD00464222 CAS Number: 4857-42-5 EINECS: 225-454-9 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...