2,4-Dichloro-N'-(2-chloroacetyl)benzenecarbohydrazide
Supplier: Apollo Scientific
OR12462-10G
OR12462-1G
OR12462-5G
APOSOR12462-10GEA
1350
EUR
APOSOR12462-10G
APOSOR12462-1G
APOSOR12462-5G
2,4-Dichloro-N'-(2-chloroacetyl)benzenecarbohydrazide
2,4-Dichloro-N'-(2-chloroacetyl)benzenecarbohydrazide
Formula:
C₉H₇Cl₃N₂O₂ MW: 281,53 g/mol Melting Pt: 204…206 °C Storage Temperature: Ambient |
MDL Number:
MFCD02580953 CAS Number: 184877-74-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...