1-(4-Methoxyphenyl)-2-oxo-3-pyrrolidinecarbonitrile

Supplier: Apollo Scientific

OR12470-1G OR12470-500MG
APOSOR12470-1GEA 270 EUR
APOSOR12470-1G APOSOR12470-500MG
1-(4-Methoxyphenyl)-2-oxo-3-pyrrolidinecarbonitrile
1-(4-Methoxyphenyl)-2-oxo-3-pyrrolidinecarbonitrile
Formula: C₁₂H₁₂N₂O₂
MW: 216,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09817457
CAS Number: 930298-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR