4-Chloro-N-methylbenzenesulphonamide

Supplier: Apollo Scientific
Warning

OR12483-1G OR12483-5G
APOSOR12483-1GEA 121 EUR
APOSOR12483-1G APOSOR12483-5G
4-Chloro-N-methylbenzenesulphonamide
4-Chloro-N-methylbenzenesulphonamide
Formula: C₇H₈ClNO₂S
Melting Pt: 52…54 °C
Storage Temperature: Ambient
MDL Number: MFCD00464330
CAS Number: 6333-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR