L-Azetidine-2-carboxylic acid

Supplier: Apollo Scientific
Warning

Synonyms: L-Trimethyleneimine-2-carboxylic Acid, (S)-Azetidine-2-carboxylic acid, (S)-(-)-2-Azetidinecarboxylic acid, L-Azetidine-2-carboxylic Acid [Antagonist of L-Proline]

OR1250T-1G
APOSOR1250T-1GEA 75.4 EUR
APOSOR1250T-1G
L-Azetidine-2-carboxylic acid
L-Azetidine-2-carboxylic acid

A four-membered ring analog of L-Proline used as an intermediate in the synthesis of polypeptides.

Formula: C₄H₇NO₂
MW: 101,11 g/mol
Melting Pt: 206…207 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005166
CAS Number: 2133-34-8
EINECS: 218-362-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR