4-Oxo-4,5,6,7-tetrahydro-1-benzofuran-3-carboxylic acid

Supplier: Apollo Scientific
Danger

Synonyms: 4-Oxo-4,5,6,7-tetrahydrobenzofuran-3-carboxylic acid

OR1251-1G OR1251-5G
APOSOR1251-1GEA 0 EUR
APOSOR1251-1G APOSOR1251-5G
4-Oxo-4,5,6,7-tetrahydro-1-benzofuran-3-carboxylic acid
4-Oxo-4,5,6,7-tetrahydro-1-benzofuran-3-carboxylic acid
Formula: C₉H₈O₄
MW: 180,16 g/mol
Melting Pt: 141…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00052170
CAS Number: 56671-28-4
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR