3-Phenethylpyrrolidine 95%

Supplier: Apollo Scientific
Warning

OR12551-250MG OR12551-1G
APOSOR12551-250MGEA 522 EUR
APOSOR12551-250MG APOSOR12551-1G
3-Phenethylpyrrolidine 95%
3-Phenethylpyrrolidine
Formula: C₁₂H₁₇N
Storage Temperature: Ambient
MDL Number: MFCD01862538
CAS Number: 613676-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR