o-Toluic hydrazide
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
o-Toluic acid hydrazide, 2-Methylbenzohydrazide
OR12590-25G
APOSOR12590-25GEA
94.4
EUR
APOSOR12590-25G
o-Toluic hydrazide
o-Toluic hydrazide
Formula:
C₈H₁₀N₂O MW: 150,18 g/mol Melting Pt: 122…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014756 CAS Number: 7658-80-2 EINECS: 231-623-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...