4-Phenoxybenzylamine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(4-Phenoxyphenyl)methanamine
OR12601-1G
OR12601-5G
OR12601-25G
APOSOR12601-1GEA
21.9
EUR
APOSOR12601-1G
APOSOR12601-5G
APOSOR12601-25G
4-Phenoxybenzylamine
4-Phenoxybenzylamine
Formula:
C₁₃H₁₃NO MW: 199.25 g/mol Boiling Pt: 138 °C/5mmHg Storage Temperature: Ambient |
MDL Number:
MFCD01310836 CAS Number: 107622-80-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...