4-(2-Pyrazinyl)benzenecarbaldehyde

Supplier: Apollo Scientific

Synonyms: 4-(Pyrazin-2-yl)benzaldehyde

OR12647-1G OR12647-250MG
APOSOR12647-1GEA 405 EUR
APOSOR12647-1G APOSOR12647-250MG
4-(2-Pyrazinyl)benzenecarbaldehyde
4-(2-Pyrazinyl)benzenecarbaldehyde
Formula: C₁₁H₈N₂O
MW: 184,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02684109
CAS Number: 127406-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR