4-Hydroxy-3-nitroanisole
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Methoxy-2-nitrophenol
OR12652-100G
OR12652-25G
OR12652-5G
APOSOR12652-100GEA
143
EUR
APOSOR12652-100G
APOSOR12652-25G
APOSOR12652-5G
4-Hydroxy-3-nitroanisole
4-Hydroxy-3-nitroanisole
Formula:
C₇H₇NO₄ MW: 169,14 g/mol Melting Pt: 78…81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00024247 CAS Number: 1568-70-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...