3-Methylsulphonylpyrrolidine

Supplier: Apollo Scientific
Warning

OR12678-1G OR12678-250MG
APOSOR12678-1GEA 511 EUR
APOSOR12678-1G APOSOR12678-250MG
3-Methylsulphonylpyrrolidine
3-Methylsulphonylpyrrolidine
Formula: C₅H₁₁NO₂S
MW: 149.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01862542
CAS Number: 433980-62-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR