4-Methyl-1,2,3-thiadiazole-5-carbonyl chloride 90%
Supplier: Apollo Scientific
|
Danger
|
OR1272-1G
OR1272-5G
APOSOR1272-1GEA
0
EUR
APOSOR1272-1G
APOSOR1272-5G
4-Methyl-1,2,3-thiadiazole-5-carbonyl chloride 90%
4-Methyl-1,2,3-thiadiazole-5-carbonyl chloride
Formula:
C₄H₃ClN₂OS Boiling Pt: 104…108 °C/18mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00173831 CAS Number: 59944-65-9 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...