4-Bromo-1H-indazole

Supplier: Apollo Scientific
Warning

OR12759-25G OR12759-5G
APOSOR12759-25GEA 58 EUR
APOSOR12759-25G APOSOR12759-5G
4-Bromo-1H-indazole
4-Bromo-1H-indazole
Formula: C₇H₅BrN₂
Storage Temperature: Ambient
MDL Number: MFCD05664001
CAS Number: 186407-74-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR