N,O-Dimethylhydroxylamine hydrochloride 98%
Supplier: Apollo Scientific
|
Warning
|
OR12778-100G
OR12778-1KG
APOSOR12778-100GEA
14.7
EUR
APOSOR12778-100G
APOSOR12778-1KG
N,O-Dimethylhydroxylamine hydrochloride 98%
N,O-Dimethylhydroxylamine hydrochloride
Formula:
C₂H₈ClNO MW: 97,55 g/mol Melting Pt: 112…116 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012485 CAS Number: 6638-79-5 EINECS: 229-642-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...