3-Amino-5-bromopyridin-2(1H)-one 98%

Supplier: Apollo Scientific

Synonyms: 3-Amino-5-bromo-2-hydroxypyridine

OR12788-1G OR12788-5G OR12788-25G
APOSOR12788-1GEA 59.7 EUR
APOSOR12788-1G APOSOR12788-5G APOSOR12788-25G
3-Amino-5-bromopyridin-2(1H)-one 98%
3-Amino-5-bromopyridin-2(1H)-one
Formula: C5H5BrN2O
MW: 189,01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03840438
CAS Number: 98786-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR