5-Chloroisoquinoline 95%
Supplier: Apollo Scientific
OR12855-5G
APOSOR12855-5GEA
186
EUR
APOSOR12855-5G
5-Chloroisoquinoline 95%
5-Chloroisoquinoline
Formula:
C₉H₆ClN MW: 163,6 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02683295 CAS Number: 5430-45-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...