7-Chloro-2-tetralone
Supplier: Apollo Scientific
Synonyms:
7-Chloro-3,4-dihydro-1H-naphthalen-2-one
OR12937-1G
OR12937-250MG
APOSOR12937-1GEA
329
EUR
APOSOR12937-1G
APOSOR12937-250MG
7-Chloro-2-tetralone
7-Chloro-2-tetralone
Formula:
C₁₀H₉ClO MW: 180,63 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04038575 CAS Number: 17556-19-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...