4-(4-Piperidyl)pyridine
Supplier: Apollo Scientific
|
Warning
|
OR13006-1G
OR13006-250MG
OR13006-5G
APOSOR13006-1GEA
112
EUR
APOSOR13006-1G
APOSOR13006-250MG
APOSOR13006-5G
4-(4-Piperidyl)pyridine
4-(4-Piperidyl)pyridine
Formula:
C₁₀H₁₄N₂ MW: 162,24 g/mol Melting Pt: 86…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD05863877 CAS Number: 581-45-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...