5-Bromopicolinamide 97%
Supplier: Apollo Scientific
|
Warning
|
OR13060-1G
OR13060-5G
OR13060-250MG
APOSOR13060-1GEA
0
EUR
APOSOR13060-1G
APOSOR13060-5G
APOSOR13060-250MG
5-Bromopicolinamide 97%
5-Bromopicolinamide
Formula:
C₆H₅BrN₂O Melting Pt: 218…220 °C Storage Temperature: Ambient |
MDL Number:
MFCD04066715 CAS Number: 90145-48-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...