4-Chloro-2-methoxyphenylboronic acid

Supplier: Apollo Scientific
Warning

Synonyms: 4-Chloro-2-methoxybenzeneboronic acid, (4-Chloro-2-methoxyphenyl)boronic acid

OR13111-1G OR13111-5G
APOSOR13111-1GEA 21.9 EUR
APOSOR13111-1G APOSOR13111-5G
4-Chloro-2-methoxyphenylboronic acid
4-Chloro-2-methoxyphenylboronic acid
Formula: C₇H₈BClO₃
MW: 186,40 g/mol
Melting Pt: 128…134 °C
Storage Temperature: Ambient
MDL Number: MFCD02684317
CAS Number: 762287-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR