4-Benzyloxyindole

Supplier: Apollo Scientific
Warning

OR13129-5G OR13129-25G OR13129-1G
APOSOR13129-5GEA 39.3 EUR
APOSOR13129-5G APOSOR13129-25G APOSOR13129-1G
4-Benzyloxyindole
4-Benzyloxyindole
Formula: C₁₅H₁₃NO
MW: 223.27 g/mol
Melting Pt: 57…61 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00047200
CAS Number: 20289-26-3
EINECS: 243-690-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR