3-(4-Methoxy-1-naphthyl)acrylic acid

Warning

OR13150-1G OR13150-100MG
APOSOR13150-1GEA 142 EUR
APOSOR13150-1G APOSOR13150-100MG
3-(4-Methoxy-1-naphthyl)acrylic acid
3-(4-Methoxy-1-naphthyl)acrylic acid
Formula: C₁₄H₁₂O₃
MW: 228,25 g/mol
Melting Pt: 225 °C
Storage Temperature: Ambient
MDL Number: MFCD00156955
CAS Number: 15971-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR