3-[1,1'-Biphenyl]-4-ylacrylic acid

Supplier: Apollo Scientific
Warning

Synonyms: 4-Phenylcinnamic acid

OR13178-25G
APOSOR13178-25GEA 0 EUR
APOSOR13178-25G
3-[1,1'-Biphenyl]-4-ylacrylic acid
3-[1,1'-Biphenyl]-4-ylacrylic acid
Formula: C₁₅H₁₂O₂
MW: 224,26 g/mol
Melting Pt: 223…225 °C
Storage Temperature: Ambient
MDL Number: MFCD00014010
CAS Number: 13026-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR