2-Oxaspiro[4.4]nonane-1,3-dione

Supplier: Apollo Scientific
Warning

Synonyms: 1,3-Dioxo-2-oxaspiro[4.4]nonane, 3-oxaspiro[4.4]nonane-2,4-dione, 8-oxaspiro[4.4]nonane-7,9-dione

OR1319-10G OR1319-1G OR1319-5G
APOSOR1319-10GEA 841 EUR
APOSOR1319-10G APOSOR1319-1G APOSOR1319-5G
2-Oxaspiro[4.4]nonane-1,3-dione
2-Oxaspiro[4.4]nonane-1,3-dione
Formula: C₈H₁₀O₃
Melting Pt: 26…28 °C
Storage Temperature: Ambient
MDL Number: MFCD00661793
CAS Number: 5623-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR