4-(Acetamidomethyl)benzeneboronic acid 97%

Supplier: Apollo Scientific
Warning

OR1333-250MG OR1333-100MG
APOSOR1333-250MGEA 56.7 EUR
APOSOR1333-250MG APOSOR1333-100MG
4-(Acetamidomethyl)benzeneboronic acid 97%
4-(Acetamidomethyl)benzeneboronic acid
Formula: C₉H₁₂BNO₃
Melting Pt: 278…280 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06659818
CAS Number: 850568-41-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR