5-Amino-3-methylisoxazole-4-carbonitrile 95%

Supplier: Apollo Scientific
Warning

OR13382-1G OR13382-5G OR13382-10G
APOSOR13382-1GEA 91.6 EUR
APOSOR13382-1G APOSOR13382-5G APOSOR13382-10G
5-Amino-3-methylisoxazole-4-carbonitrile 95%
5-Amino-3-methylisoxazole-4-carbonitrile
Formula: C₅H₅N₃O
Melting Pt: 227…230 °C
Storage Temperature: Ambient
MDL Number: MFCD00027378
CAS Number: 35261-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR