3-Amino-2-pyridone 97%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Amino-1,2-dihydro-2-oxopyridine, 1,2-Dihydro-2-oxopyridin-3-amine

OR13432-25G OR13432-5G
APOSOR13432-25GEA 205 EUR
APOSOR13432-25G APOSOR13432-5G
3-Amino-2-pyridone 97%
3-Aminopyridin-2(1H)-one
Formula: C₅H₆N₂O
MW: 110,12 g/mol
Melting Pt: 124 °C
Storage Temperature: Ambient
MDL Number: MFCD02752887
CAS Number: 33630-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR