1-(5-Bromopyridin-3-yl)-2-methylsulphonylethanone 95+%

Supplier: Apollo Scientific

OR1346-1G OR1346-5G OR1346-25G
APOSOR1346-1GEA 136 EUR
APOSOR1346-1G APOSOR1346-5G APOSOR1346-25G
1-(5-Bromopyridin-3-yl)-2-methylsulphonylethanone 95+%
1-(5-Bromopyridin-3-yl)-2-methylsulphonylethanone
Formula: C₈H₈BrNO₃S
MW: 278.13 g/mol
Melting Pt: 158…161 °C
Storage Temperature: Ambient
MDL Number: MFCD01941313
CAS Number: 386715-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR