4-Ethylbenzenesulphonamide 98%

Supplier: Apollo Scientific
Warning

OR1356-100G OR1356-25G OR1356-5G
APOSOR1356-100GEA 58 EUR
APOSOR1356-100G APOSOR1356-25G APOSOR1356-5G
4-Ethylbenzenesulphonamide 98%
4-Ethylbenzenesulphonamide
Formula: C₈H₁₁NO₂S
MW: 185.24 g/mol
Melting Pt: 111 °C
Storage Temperature: Ambient
MDL Number: MFCD01365828
CAS Number: 138-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR