2-Nitro-1-phenyl-1-ethanol

Supplier: Apollo Scientific
Warning

Synonyms: 2-Nitro-1-phenylethanol

OR13560-1G OR13560-500MG
APOSOR13560-1GEA 858 EUR
APOSOR13560-1G APOSOR13560-500MG
2-Nitro-1-phenyl-1-ethanol
2-Nitro-1-phenyl-1-ethanol
Formula: C₈H₉NO₃
MW: 167,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00854219
CAS Number: 15990-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR