2,4,6-Trimethylbenzenesulphonamide

Supplier: Apollo Scientific
Warning

OR1357-25G OR1357-5G OR1357-100G
APOSOR1357-25GEA 16.1 EUR
APOSOR1357-25G APOSOR1357-5G APOSOR1357-100G
2,4,6-Trimethylbenzenesulphonamide
2,4,6-Trimethylbenzenesulphonamide
Formula: C₉H₁₃NO₂S
MW: 199,2700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00463245
CAS Number: 4543-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR