5-Bromo-7-methyl-1H-indazole

Supplier: Apollo Scientific
Warning

OR13583-1G OR13583-5G OR13583-250MG
APOSOR13583-1GEA 16.1 EUR
APOSOR13583-1G APOSOR13583-5G APOSOR13583-250MG
5-Bromo-7-methyl-1H-indazole
5-Bromo-7-methyl-1H-indazole
Formula: C₈H₇BrN₂
MW: 211,0600 g/mol
Melting Pt: 185…187 °C
Storage Temperature: Ambient
MDL Number: MFCD03990484
CAS Number: 156454-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR