4-Hydroxy-3-pyridinesulphonic acid 97%

Supplier: Apollo Scientific
Danger

Synonyms: 4-Hydroxypyridine-3-sulphonic acid

OR13620-25G OR13620-5G OR13620-10G
APOSOR13620-25GEA 81.2 EUR
APOSOR13620-25G APOSOR13620-5G APOSOR13620-10G
4-Hydroxy-3-pyridinesulphonic acid 97%
4-Hydroxy-3-pyridinesulphonic acid
Formula: C₅H₅NO₄S
MW: 175.1625 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD02093473
CAS Number: 51498-37-4
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR