Ethyl-4H-thieno[3,2-b]pyrrole-5-carboxylate

Supplier: Apollo Scientific
Warning

Synonyms: Ethyl 4H-thieno[3,2-b]pyrrole-5-carboxylate

OR13647-1G OR13647-5G OR13647-10G
APOSOR13647-1GEA 74.2 EUR
APOSOR13647-1G APOSOR13647-5G APOSOR13647-10G
Ethyl-4H-thieno[3,2-b]pyrrole-5-carboxylate
Ethyl-4H-thieno[3,2-b]pyrrole-5-carboxylate
Formula: C₉H₉NO₂S
MW: 195,2383 g/mol
Melting Pt: 131…132 °C
Storage Temperature: Ambient
MDL Number: MFCD03425800
CAS Number: 46193-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR