3-Isopropylbenzoic acid 98%

Supplier: Apollo Scientific
Warning

OR13714-5G
APOSOR13714-5GEA 120 EUR
APOSOR13714-5G
3-Isopropylbenzoic acid 98%
3-Isopropylbenzoic acid
Formula: C10H12O2
MW: 164,2011 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09038906
CAS Number: 5651-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR