2-Bromo-3-chlorothiophene 96%
Supplier: Apollo Scientific
|
Warning
|
OR13719-1G
OR13719-5G
APOSOR13719-1GEA
108
EUR
APOSOR13719-1G
APOSOR13719-5G
2-Bromo-3-chlorothiophene 96%
2-Bromo-3-chlorothiophene
Formula:
C₄H₂BrClS Boiling Pt: 73…75 °C/12mmHg Storage Temperature: Refrigerator |
MDL Number:
MFCD00043882 CAS Number: 77893-68-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...