5-Ethoxy-1H-indole 95%

Supplier: Apollo Scientific
Warning

OR13724-1G OR13724-5G
APOSOR13724-1GEA 82.8 EUR
APOSOR13724-1G APOSOR13724-5G
5-Ethoxy-1H-indole 95%
5-Ethoxy-1H-indole
Formula: C10H11NO
MW: 161.2004 g/mol
Boiling Pt: 164 °C/11mmHg
Melting Pt: 144…146 °C
Storage Temperature: Ambient
MDL Number: MFCD07364533
CAS Number: 10501-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR