4-Bromoanthranilic acid 95%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2-Amino-4-bromobenzoic acid
OR13732-100G
OR13732-25G
OR13732-5G
APOSOR13732-100GEA
66.7
EUR
APOSOR13732-100G
APOSOR13732-25G
APOSOR13732-5G
4-Bromoanthranilic acid 95%
4-Bromoanthranilic acid
Formula:
C₇H₆BrNO₂ MW: 216,03 g/mol Melting Pt: 230…234 °C Storage Temperature: Ambient |
MDL Number:
MFCD03618454 CAS Number: 20776-50-5 EINECS: 244-025-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...