2-Thiophenesulphonamide

Supplier: Apollo Scientific

Synonyms: Thiophene-2-sulphonamide

OR13858-10G OR13858-1G OR13858-5G
APOSOR13858-10GEA 65.5 EUR
APOSOR13858-10G APOSOR13858-1G APOSOR13858-5G
2-Thiophenesulphonamide
2-Thiophenesulphonamide
Formula: C₄H₅NO₂S₂
MW: 163,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00185853
CAS Number: 6339-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR