(1-Ethoxyethylidene)malononitrile
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-(1-Ethoxyethylidene)malononitrile, 2-(1-Ethoxy-ethylidene)-malononitrile
OR13891-100G
OR13891-25G
APOSOR13891-100GEA
216
EUR
APOSOR13891-100G
APOSOR13891-25G
(1-Ethoxyethylidene)malononitrile
(1-Ethoxyethylidene)malononitrile
Formula:
C₇H₈N₂O MW: 136,15 g/mol Melting Pt: 89…93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001852 CAS Number: 5417-82-3 EINECS: 226-521-5 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...