1,2-Dichloro-3-nitrobenzene

Supplier: Apollo Scientific
Warning

Synonyms: 2,3-Dichloronitrobenzene

OR1391-100G OR1391-500G
APOSOR1391-100GEA 109 EUR
APOSOR1391-100G APOSOR1391-500G
1,2-Dichloro-3-nitrobenzene
1,2-Dichloro-3-nitrobenzene
Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
Boiling Pt: 257…258 °C
Melting Pt: 61…64 °C
Density: 1,45 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007062
CAS Number: 3209-22-1
UN: 3077
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR