2-Morpholino-5-pyridineboronic acid pinacol ester
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
6-(4-Morpholinyl)pyridine-3-boronic acid pinacol ester, 2-(Morpholin-4-yl)pyridine-5-boronic acid pinacol ester, 4-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)morpholine
OR14006-1G
OR14006-5G
APOSOR14006-1GEA
64
EUR
APOSOR14006-1G
APOSOR14006-5G
2-Morpholino-5-pyridineboronic acid pinacol ester
2-Morpholino-5-pyridineboronic acid pinacol ester
Formula:
C₁₅H₂₃BN₂O₃ MW: 290,17 g/mol Melting Pt: 132…139 °C Storage Temperature: Ambient |
MDL Number:
MFCD04039874 CAS Number: 485799-04-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...