2-(3-Bromo-2-thienyl)-2-oxoethyl thiocyanate
Supplier: Apollo Scientific
OR14037-1G
OR14037-5G
APOSOR14037-1GEA
538
EUR
APOSOR14037-1G
APOSOR14037-5G
2-(3-Bromo-2-thienyl)-2-oxoethyl thiocyanate
2-(3-Bromo-2-thienyl)-2-oxoethyl thiocyanate
Formula:
C₇H₄BrNOS₂ Melting Pt: 99…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD09027130 CAS Number: 937602-37-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...