2-(4-Hydroxy-2-phenyl-1,3-thiazol-5-yl)acetic acid 95%

Supplier: Apollo Scientific
Warning

OR14126-1G OR14126-10G OR14126-5G
APOSOR14126-1GEA 149 EUR
APOSOR14126-1G APOSOR14126-10G APOSOR14126-5G
2-(4-Hydroxy-2-phenyl-1,3-thiazol-5-yl)acetic acid 95%
2-(4-Hydroxy-2-phenyl-1,3-thiazol-5-yl)acetic acid
Formula: C₁₁H₉NO₃S
MW: 235,26 g/mol
Melting Pt: 196…200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD08689717
CAS Number: 133834-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR