1-(Methoxycarbonyl)-4-piperidinecarboxylic acid

Supplier: Apollo Scientific
Warning

OR14129-1G OR14129-5G
APOSOR14129-1GEA 155 EUR
APOSOR14129-1G APOSOR14129-5G
1-(Methoxycarbonyl)-4-piperidinecarboxylic acid
1-(Methoxycarbonyl)-4-piperidinecarboxylic acid
Formula: C₈H₁₃NO₄
MW: 187,2 g/mol
Melting Pt: 97…99 °C
Storage Temperature: Ambient
MDL Number: MFCD02082494
CAS Number: 197585-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR